Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd2ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008583.2 | NP_001008583.2 | 15 - 16 | 1824 | 99 | |
XM_005160684.4 | XP_005160741.1 | 14 - 15 | 1455 | 99 | ||
XM_009294709.3 | XP_009292984.1 | 15 - 16 | 1849 | 99 | ||
GenBank mRNA | DT075242.1 | - | 4 - 5 | 421 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |