Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd2ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008583.2 | NP_001008583.2 | 2 - 3 | 412 | 67 | |
XM_009294709.3 | XP_009292984.1 | 2 - 3 | 443 | 67 | ||
GenBank mRNA | BC086846.1 | - | 1 - 2 | 271 | 67 | |
BC164631.1 | - | 1 - 2 | 82 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |