Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbm39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012304.1 | NP_001012304.1 | 14 - 15 | 1655 | 81 | |
XM_005169275.3 | XP_005169332.1 | 15 - 16 | 1641 | 81 | ||
XM_005169277.4 | XP_005169334.1 | 15 - 16 | 1732 | 81 | ||
XM_005169278.4 | XP_005169335.1 | 16 - 17 | 1700 | 81 | ||
XM_009306024.3 | XP_009304299.1 | 16 - 17 | 1931 | 81 | ||
XM_021479585.1 | XP_021335260.1 | 15 - 16 | 2202 | 81 | ||
GenBank mRNA | BC044487.1 | - | 14 - 15 | 1655 | 81 | |
BC063981.1 | - | 13 - 14 | 81 | |||
BC097076.1 | - | 16 - 17 | 1878 | 81 | ||
BC165689.1 | - | 13 - 14 | 1424 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |