Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013291.3 | NP_001013309.2 | 4 - 5 | 647 | 126 | |
GenBank mRNA | BC090282.1 | - | 4 - 5 | 651 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |