Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045373.2 | NP_001038838.2 | 4 - 5 | 312 | 66 | |
XM_017352825.2 | XP_017208314.1 | 4 - 5 | 336 | 66 | ||
XM_021468909.1 | XP_021324584.1 | 4 - 5 | 287 | 66 | ||
GenBank mRNA | BC164588.1 | - | 3 - 4 | 279 | 66 | |
EH585180.1 | - | 4 - 5 | 304 | 66 |
Gene Symbol: LOC110438496 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021471041.1 | XP_021326716.1 | 1 - 2 | 160 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Danio rerio Probe spans exons |