Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076632.1 | NP_001070100.1 | 3 - 4 | 190 | 86 | |
GenBank mRNA | BC124342.1 | - | 3 - 4 | 190 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |