Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nt5c2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079966.2 | NP_001073435.2 | 13 - 14 | 1188 | 69 | |
XM_005168392.4 | XP_005168449.1 | 13 - 14 | 1357 | 69 | ||
XM_021474992.1 | XP_021330667.1 | 12 - 13 | 1032 | 69 | ||
XM_021474993.1 | XP_021330668.1 | 8 - 9 | 550 | 69 | ||
GenBank mRNA | BC076085.1 | - | 13 - 14 | 1287 | 69 | |
BC095068.1 | - | 13 - 14 | 1439 | 69 | ||
BC124713.1 | - | 13 - 14 | 1133 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |