Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mab21l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110025.1 | NP_001103495.1 | 4 - 5 | 688 | 92 | |
XM_021478689.1 | XP_021334364.1 | 4 - 5 | 773 | 92 | ||
GenBank mRNA | BC153314.1 | - | 4 - 5 | 688 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |