Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194373.1 | NP_919354.1 | 3 - 4 | 208 | 79 | |
GenBank mRNA | AB074154.1 | - | 3 - 4 | 208 | 79 | |
BC052140.1 | - | 3 - 4 | 229 | 79 | ||
BC085553.1 | - | 3 - 4 | 249 | 79 | ||
BC164860.1 | - | 3 - 4 | 220 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |