Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199558.2 | NP_955852.2 | 14 - 15 | 2537 | 68 | |
GenBank mRNA | BC059198.1 | - | 14 - 15 | 2514 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |