Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: faua | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200737.1 | NP_957031.1 | 1 - 2 | 94 | 82 | |
GenBank mRNA | BC059522.1 | - | 1 - 2 | 94 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |