Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201206.2 | NP_957500.2 | 8 - 9 | 1208 | 71 | |
GenBank mRNA | BC046017.1 | - | 8 - 9 | 1195 | 71 | |
BC066452.1 | - | 8 - 9 | 1185 | 71 | ||
FP111012.1 | - | 1 - 2 | 471 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |