Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213151.1 | NP_998316.1 | 4 - 5 | 407 | 69 | |
GenBank mRNA | BC062383.1 | - | 4 - 5 | 407 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |