Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hspa4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214716.1 | NP_999881.1 | 10 - 11 | 1424 | 77 | |
GenBank mRNA | BC051152.1 | - | 10 - 11 | 1424 | 77 | |
BC165369.1 | - | 10 - 11 | 1274 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |