Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acp5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214773.1 | NP_999938.1 | 3 - 4 | 335 | 94 | |
XM_005165725.4 | XP_005165782.1 | 3 - 4 | 343 | 94 | ||
XM_005165726.4 | XP_005165783.1 | 3 - 4 | 420 | 94 | ||
GenBank mRNA | BC055256.1 | - | 3 - 4 | 335 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |