Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hormad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002357.1 | NP_001002357.1 | 13 - 14 | 1085 | 92 | |
XM_021466675.1 | XP_021322350.1 | 11 - 12 | 1692 | 92 | ||
GenBank mRNA | BC075898.1 | - | 13 - 14 | 1085 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |