Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam83fb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007320.3 | NP_001007321.2 | 3 - 4 | 694 | 138 | |
GenBank mRNA | BC085639.1 | - | 3 - 4 | 651 | 138 | |
EH506757.1 | - | 3 - 4 | 696 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |