Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012486.1 | NP_001012504.1 | 27 - 28 | 3753 | 64 | |
XM_009306437.3 | XP_009304712.1 | 27 - 28 | 4152 | 64 | ||
XM_009306438.3 | XP_009304713.1 | 26 - 27 | 4146 | 64 | ||
XM_009306439.3 | XP_009304714.1 | 27 - 28 | 4152 | 64 | ||
XM_009306440.3 | XP_009304715.1 | 27 - 28 | 4152 | 64 | ||
GenBank mRNA | AY772391.1 | - | 27 - 28 | 3753 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |