Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sema6dl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017682.2 | NP_001017682.2 | 14 - 15 | 1669 | 63 | |
XM_005173648.4 | XP_005173705.1 | 14 - 15 | 1787 | 63 | ||
XM_005173649.4 | XP_005173706.1 | 14 - 15 | 1787 | 63 | ||
XM_005173650.4 | XP_005173707.1 | 14 - 15 | 1787 | 63 | ||
XM_009293805.3 | XP_009292080.1 | 14 - 15 | 2178 | 63 | ||
XM_009293807.3 | XP_009292082.1 | 14 - 15 | 1787 | 63 | ||
GenBank mRNA | AB212073.1 | - | 14 - 15 | 1669 | 63 | |
AB212074.1 | - | 14 - 15 | 1669 | 63 | ||
CU458858.1 | - | 13 - 14 | 1575 | 63 | ||
CU458860.1 | - | 13 - 14 | 1569 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons |