Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gmpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020612.1 | NP_001018448.1 | 4 - 5 | 537 | 107 | |
GenBank mRNA | BC095337.1 | - | 4 - 5 | 537 | 107 | |
BC164152.1 | - | 4 - 5 | 502 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |