Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ela3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024408.1 | NP_001019579.1 | 4 - 5 | 365 | 83 | |
XM_005171867.4 | XP_005171924.1 | 4 - 5 | 433 | 83 | ||
XM_021476165.1 | XP_021331840.1 | 4 - 5 | 433 | 83 | ||
GenBank mRNA | AY583322.1 | - | 4 - 5 | 361 | 83 | |
BC095070.1 | - | 4 - 5 | 365 | 83 | ||
BC165005.1 | - | 2 - 3 | 188 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |