Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myh11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024448.1 | NP_001019619.1 | 37 - 38 | 5395 | 110 | |
XM_021476875.1 | XP_021332550.1 | 38 - 39 | 5448 | 110 | ||
XM_021476876.1 | XP_021332551.1 | 38 - 39 | 5448 | 110 | ||
GenBank mRNA | BC162152.1 | - | 37 - 38 | 5359 | 110 | |
DQ022376.1 | - | 37 - 38 | 5395 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |