Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024819.1 | NP_001019990.1 | 7 - 8 | 1133 | 70 | |
GenBank mRNA | AY613958.1 | - | 7 - 8 | 1133 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |