Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077178.2 | NP_001070646.1 | 8 - 9 | 1194 | 77 | |
XM_005157588.4 | XP_005157645.1 | 8 - 9 | 995 | 77 | ||
XM_009291665.3 | XP_009289940.1 | 7 - 8 | 1172 | 77 | ||
GenBank mRNA | BC124265.1 | - | 8 - 9 | 1195 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |