Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zc3hc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077378.1 | NP_001070846.1 | 5 - 6 | 654 | 106 | |
GenBank mRNA | BC124810.1 | - | 5 - 6 | 654 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |