Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacnb2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081748.2 | NP_001075217.2 | 820 | 92 | ||
NM_001166324.1 | NP_001159796.1 | 578 | 92 | |||
GenBank mRNA | BC160620.1 | - | 5 - 6 | 469 | 92 | |
BC162962.1 | - | 5 - 6 | 436 | 92 | ||
BC162994.1 | - | 5 - 6 | 436 | 92 | ||
DQ372944.1 | - | 5 - 6 | 469 | 92 | ||
EU301442.1 | - | 820 | 92 | |||
EU301443.1 | - | 578 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |