Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082866.1 | NP_001076335.1 | 6 - 7 | 784 | 65 | |
GenBank mRNA | BC134140.1 | - | 6 - 7 | 784 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |