Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fcer1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100106.2 | NP_001093576.2 | 4 - 5 | 267 | 66 | |
GenBank mRNA | EF158447.1 | - | 4 - 5 | 280 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Danio rerio Inventoried Probe spans exons |