Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drosha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110472.1 | NP_001103942.1 | 5 - 6 | 1428 | 65 | |
GenBank mRNA | BC133134.1 | - | 5 - 6 | 1428 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |