Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: erh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271817.1 | NP_001258746.1 | 3 - 4 | 342 | 110 | |
GenBank mRNA | BC059528.1 | - | 3 - 4 | 262 | 110 | |
CT674675.2 | - | 1 - 2 | 611 | 110 | ||
EH478279.1 | - | 1 - 2 | 588 | 110 | ||
EH543119.1 | - | 3 - 4 | 376 | 110 | ||
U66872.1 | - | 2 - 3 | 211 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |