Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mapk12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131407.1 | NP_571482.1 | 9 - 10 | 883 | 163 | |
GenBank mRNA | BC085415.1 | - | 9 - 10 | 883 | 163 | |
BC165821.1 | - | 9 - 10 | 801 | 163 | ||
DQ360071.1 | - | 9 - 10 | 767 | 163 | ||
Y15075.1 | - | 851 | 163 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |