Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144763.1 | NP_658909.1 | 2 - 3 | 304 | 141 | |
GenBank mRNA | AJ306395.1 | - | 2 - 3 | 304 | 141 | |
BC044450.1 | - | 2 - 3 | 290 | 141 | ||
BC083507.1 | - | 2 - 3 | 288 | 141 | ||
BC165764.1 | - | 2 - 3 | 245 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |