Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rock2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174863.2 | NP_777288.1 | 21 - 22 | 2958 | 140 | |
GenBank mRNA | AF295804.1 | - | 21 - 22 | 2958 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |