Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198807.1 | NP_942102.1 | 12 - 13 | 1932 | 122 | |
GenBank mRNA | BC045956.1 | - | 12 - 13 | 1932 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |