Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198807.1 | NP_942102.1 | 7 - 8 | 1257 | 86 | |
GenBank mRNA | BC045956.1 | - | 7 - 8 | 1257 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |