Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttc9c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200265.1 | NP_956559.1 | 2 - 3 | 390 | 106 | |
GenBank mRNA | BC049032.1 | - | 2 - 3 | 390 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |