Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ociad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200302.2 | NP_956596.1 | 2 - 3 | 243 | 81 | |
GenBank mRNA | BC049514.1 | - | 2 - 3 | 245 | 81 | |
BC066633.1 | - | 2 - 3 | 279 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |