Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc22a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200849.1 | NP_957143.1 | 8 - 9 | 1464 | 77 | |
XM_021468778.1 | XP_021324453.1 | 8 - 9 | 3145 | 77 | ||
GenBank mRNA | BC062384.1 | - | 8 - 9 | 1464 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |