Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nup50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201580.2 | NP_963874.2 | 6 - 7 | 1065 | 83 | |
GenBank mRNA | AY398390.1 | - | 6 - 7 | 1064 | 83 | |
BC051604.1 | - | 6 - 7 | 1013 | 83 | ||
BC065637.1 | - | 6 - 7 | 1065 | 83 | ||
BC153634.1 | - | 4 - 5 | 737 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |