Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ybey | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328511.1 | NP_001315440.1 | 2 - 3 | 501 | 75 | |
NM_212594.2 | NP_997759.1 | 2 - 3 | 501 | 75 | ||
GenBank mRNA | BC067584.1 | - | 2 - 3 | 471 | 75 | |
EH436338.1 | - | 2 - 3 | 525 | 75 | ||
EH587575.1 | - | 2 - 3 | 525 | 75 |
Gene Symbol: LOC110438491 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021471013.1 | XP_021326688.1 | 2 - 3 | 504 | 75 | |
XM_021471014.1 | XP_021326689.1 | 2 - 3 | 504 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |