Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnase1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002674.1 | NP_001002674.1 | 3 - 4 | 320 | 82 | |
GenBank mRNA | BC076479.1 | - | 3 - 4 | 320 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |