Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sept5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003782.2 | NP_001003782.1 | 8 - 9 | 727 | 114 | |
XM_021479157.1 | XP_021334832.1 | 8 - 9 | 792 | 114 | ||
XM_021479158.1 | XP_021334833.1 | 7 - 8 | 705 | 114 | ||
GenBank mRNA | BC078256.1 | - | 8 - 9 | 731 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |