Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007352.1 | NP_001007353.1 | 5 - 6 | 641 | 73 | |
XM_005160065.4 | XP_005160122.1 | 6 - 7 | 734 | 73 | ||
GenBank mRNA | BC085525.1 | - | 5 - 6 | 641 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |