Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017681.1 | NP_001017681.1 | 5 - 6 | 604 | 102 | |
GenBank mRNA | BC093160.1 | - | 5 - 6 | 604 | 102 | |
BC165257.1 | - | 5 - 6 | 481 | 102 |
Gene Symbol: LOC100536377 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003201379.4 | XP_003201427.1 | 5 - 6 | 627 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |