Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fstl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017860.1 | NP_001017860.1 | 8 - 9 | 752 | 70 | |
GenBank mRNA | BC092990.1 | - | 8 - 9 | 752 | 70 | |
DQ317970.1 | - | 8 - 9 | 745 | 70 | ||
KM655580.1 | - | 7 - 8 | 696 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |