Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kmt5aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045349.2 | NP_001038814.2 | 4 - 5 | 592 | 79 | |
GenBank mRNA | BC122396.1 | - | 4 - 5 | 618 | 79 | |
BC165703.1 | - | 3 - 4 | 431 | 79 | ||
DQ343297.1 | - | 4 - 5 | 592 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |