Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dok6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089439.1 | NP_001082908.1 | 7 - 8 | 970 | 69 | |
XM_009297335.3 | XP_009295610.1 | 5 - 6 | 637 | 69 | ||
GenBank mRNA | BC133919.1 | - | 7 - 8 | 970 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |