Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rragd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099424.2 | NP_001092894.2 | 3 - 4 | 857 | 117 | |
XM_021467215.1 | XP_021322890.1 | 3 - 4 | 883 | 117 | ||
GenBank mRNA | BC142594.1 | - | 3 - 4 | 857 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |