Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131191.1 | NP_571266.1 | 2 - 3 | 197 | 83 | |
GenBank mRNA | BC060942.1 | - | 2 - 3 | 184 | 83 | |
Y17256.1 | - | 197 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |