Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rxrbb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131238.1 | NP_571313.1 | 10 - 11 | 1232 | 63 | |
XM_021466616.1 | XP_021322291.1 | 10 - 11 | 1209 | 63 | ||
XM_021466617.1 | XP_021322292.1 | 10 - 11 | 1211 | 63 | ||
GenBank mRNA | BC162301.1 | - | 10 - 11 | 1226 | 63 | |
BC162302.1 | - | 10 - 11 | 1226 | 63 | ||
U29941.1 | - | 10 - 11 | 1232 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |